Assay Details
| Gene Symbol: Rsad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013381.2 | NP_001013399.1 | 2 - 3 | 352 | 115 | |
| GenBank mRNA | AK046706.1 | - | 2 - 3 | 357 | 115 | |
| AK148401.1 | - | 2 - 3 | 347 | 115 | ||
| BB645699.1 | - | 2 - 3 | 357 | 115 | ||
| BC139274.1 | - | 2 - 3 | 295 | 8862296 | 115 | |
| BC139276.1 | - | 2 - 3 | 295 | 8862298 | 115 | |
| BC145594.1 | - | 2 - 3 | 295 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rsad1
Genomic Map