Assay Details
| Gene Symbol: Xylt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145828.3 | NP_665827.2 | 4 - 5 | 1089 | 94 | |
| GenBank mRNA | AJ291751.2 | - | 4 - 5 | 1006 | 94 | |
| AK083482.1 | - | 4 - 5 | 1058 | 94 | ||
| AK086804.1 | - | 4 - 5 | 1021 | 94 | ||
| AK150521.1 | - | 4 - 5 | 1042 | 94 | ||
| AK153332.1 | - | 4 - 5 | 1042 | 94 | ||
| AK158056.1 | - | 4 - 5 | 1089 | 94 | ||
| BC034082.1 | - | 3 - 4 | 608 | 4239646 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xylt2
Genomic Map