| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300008938.mm.1 |
NM_023186.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008938.mm.1 |
NM_023186.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008938.mm.1 |
TC0300001102.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300008935.mm.1 |
NM_023186.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008935.mm.1 |
NM_023186.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008935.mm.1 |
TC0300001102.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300008939.mm.1 |
NM_023186.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008939.mm.1 |
NM_023186.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008939.mm.1 |
TC0300001102.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300008937.mm.1 |
NM_023186.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008937.mm.1 |
NM_023186.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300008937.mm.1 |
TC0300001102.mm.1 |
MTA TC |