Assay Details
| Gene Symbol: Tmigd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027025.4 | NP_081301.2 | 5 - 6 | 770 | 65 | |
| GenBank mRNA | AK005581.1 | - | 5 - 6 | 722 | 65 | |
| BC038878.1 | - | 4 - 5 | 650 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmigd3
Genomic Map