Assay Details
| Gene Symbol: Fmnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011711.1 | NP_035841.1 | 6 - 7 | 775 | 73 | |
| GenBank mRNA | AK129473.1 | - | 6 - 7 | 694 | 73 | |
| AK139711.1 | - | 6 - 7 | 758 | 73 | ||
| AK143308.1 | - | 6 - 7 | 674 | 73 | ||
| AK155331.1 | - | 6 - 7 | 792 | 73 | ||
| BC023797.1 | - | 5 - 6 | 495 | 73 | ||
| BC085094.1 | - | 6 - 7 | 672 | 73 | ||
| BC131961.1 | - | 6 - 7 | 651 | 40130238 | 73 | |
More Information
Set Membership: |
|

Fmnl3
Genomic Map