Assay Details
| Gene Symbol: Adcy6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007405.2 | NP_031431.2 | 14 - 15 | 2436 | 74 | |
| GenBank mRNA | AK122279.1 | - | 1 - 2 | 82 | 74 | |
| AK147516.1 | - | 14 - 15 | 2436 | 74 | ||
| BC145101.1 | - | 13 - 14 | 2269 | 74 | ||
| M93422.1 | - | 13 - 14 | 2368 | 74 | ||
| M96653.1 | - | 13 - 14 | 2258 | 74 | ||
More Information
Set Membership: |
|

Adcy6
Genomic Map