Assay Details
Gene Symbol: 1700125H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028589.1 | NP_082865.1 | 4 - 5 | 517 | 58 | |
GenBank mRNA | AK007281.1 | - | 3 - 4 | 239 | 58 | |
BC048674.1 | - | 4 - 5 | 517 | 6774125 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
