Assay Details
| Gene Symbol: Appbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025825.3 | NP_080101.1 | 7 - 8 | 1142 | 107 | |
| GenBank mRNA | AK004888.1 | - | 7 - 8 | 1153 | 107 | |
| AK005440.1 | - | 7 - 8 | 848 | 107 | ||
| AK122222.1 | - | 749 | 107 | |||
| AK133853.1 | - | 7 - 8 | 1134 | 107 | ||
| BC018442.1 | - | 7 - 8 | 1120 | 3157290 | 107 | |
| BC019152.1 | - | 7 - 8 | 1143 | 5065139 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Appbp2
Genomic Map