Assay Details
| Gene Symbol: Appbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025825.3 | NP_080101.1 | 8 - 9 | 1248 | 104 | |
| GenBank mRNA | AK005440.1 | - | 8 - 9 | 954 | 104 | |
| AK122222.1 | - | 855 | 104 | |||
| AK133853.1 | - | 8 - 9 | 1240 | 104 | ||
| BC018442.1 | - | 8 - 9 | 1226 | 3157290 | 104 | |
| BC019152.1 | - | 8 - 9 | 1249 | 5065139 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Appbp2
Genomic Map