Assay Details
| Gene Symbol: Elovl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029001.5 | NP_083277.3 | 3 - 4 | 424 | 125 | |
| GenBank mRNA | AK018616.1 | - | 3 - 4 | 399 | 125 | |
| AK037162.1 | - | 3 - 4 | 396 | 125 | ||
| AK049118.1 | - | 3 - 4 | 424 | 125 | ||
| AK137668.1 | - | 3 - 4 | 409 | 125 | ||
| BC005602.1 | - | 3 - 4 | 424 | 3483017 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elovl7
Genomic Map