Assay Details
Gene Symbol: Elovl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029001.5 | NP_083277.3 | 5 - 6 | 549 | 93 | |
GenBank mRNA | AK018616.1 | - | 5 - 6 | 524 | 93 | |
AK037162.1 | - | 5 - 6 | 521 | 93 | ||
AK049118.1 | - | 5 - 6 | 549 | 93 | ||
AK050441.1 | - | 5 - 6 | 498 | 93 | ||
AK137668.1 | - | 5 - 6 | 534 | 93 | ||
BC005602.1 | - | 5 - 6 | 549 | 3483017 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
