Assay Details
| Gene Symbol: Ahsa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146036.1 | NP_666148.1 | 3 - 4 | 485 | 96 | |
| GenBank mRNA | AK076069.1 | - | 3 - 4 | 495 | 96 | |
| AK165289.1 | - | 3 - 4 | 505 | 96 | ||
| AK166644.1 | - | 3 - 4 | 458 | 96 | ||
| AK201587.1 | - | 3 - 4 | 299 | 96 | ||
| BC023857.1 | - | 3 - 4 | 452 | 5346745 | 96 | |
| BC025552.1 | - | 3 - 4 | 485 | 5320590 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ahsa1
Genomic Map