Assay Details
| Gene Symbol: Adsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009634.6 | NP_033764.2 | 3 - 4 | 464 | 109 | |
| GenBank mRNA | AK049372.1 | - | 3 - 4 | 436 | 109 | |
| AK051111.1 | - | 3 - 4 | 436 | 109 | ||
| AK152092.1 | - | 3 - 4 | 436 | 109 | ||
| AK166019.1 | - | 3 - 4 | 432 | 109 | ||
| AK168906.1 | - | 3 - 4 | 429 | 109 | ||
| AK171725.1 | - | 3 - 4 | 445 | 109 | ||
| BC020187.1 | - | 3 - 4 | 440 | 4913637 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adsl
Genomic Map