Assay Details
| Gene Symbol: Adsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009634.6 | NP_033764.2 | 6 - 7 | 755 | 68 | |
| GenBank mRNA | AK049372.1 | - | 6 - 7 | 727 | 68 | |
| AK051111.1 | - | 6 - 7 | 727 | 68 | ||
| AK152092.1 | - | 6 - 7 | 727 | 68 | ||
| AK166019.1 | - | 6 - 7 | 723 | 68 | ||
| AK168906.1 | - | 6 - 7 | 720 | 68 | ||
| AK170216.1 | - | 5 - 6 | 640 | 68 | ||
| AK171725.1 | - | 6 - 7 | 736 | 68 | ||
| BC020187.1 | - | 6 - 7 | 731 | 4913637 | 68 | |
| U20225.1 | - | 6 - 7 | 731 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adsl
Genomic Map