Assay Details
| Gene Symbol: Adsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009634.6 | NP_033764.2 | 8 - 9 | 924 | 88 | |
| GenBank mRNA | AK049372.1 | - | 8 - 9 | 896 | 88 | |
| AK051111.1 | - | 8 - 9 | 896 | 88 | ||
| AK152092.1 | - | 8 - 9 | 896 | 88 | ||
| AK166019.1 | - | 8 - 9 | 892 | 88 | ||
| AK168906.1 | - | 8 - 9 | 889 | 88 | ||
| AK171725.1 | - | 8 - 9 | 905 | 88 | ||
| BC020187.1 | - | 8 - 9 | 900 | 4913637 | 88 | |
| U20225.1 | - | 8 - 9 | 900 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adsl
Genomic Map