Assay Details
| Gene Symbol: Gramd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205353.1 | NP_001192282.1 | 3 - 4 | 567 | 73 | |
| NM_172611.4 | NP_766199.1 | 3 - 4 | 469 | 73 | ||
| GenBank mRNA | AK036832.1 | - | 3 - 4 | 458 | 73 | |
| AK129209.1 | - | 3 - 4 | 371 | 73 | ||
| AK142266.1 | - | 3 - 4 | 422 | 73 | ||
| BC085148.1 | - | 3 - 4 | 525 | 73 | ||
| BC139228.1 | - | 3 - 4 | 458 | 8862250 | 73 | |
| BC139229.1 | - | 3 - 4 | 458 | 8862251 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gramd4
Genomic Map