Assay Details
Gene Symbol: Dhx29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172594.2 | NP_766182.2 | 24 - 25 | 3814 | 85 | |
GenBank mRNA | AK028342.1 | - | 12 - 13 | 1584 | 85 | |
BC057112.1 | - | 24 - 25 | 3887 | 6417650 | 85 | |
BC082319.1 | - | 24 - 25 | 3877 | 6417162 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
