Assay Details
| Gene Symbol: Il6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010560.3 | NP_034690.3 | 8 - 9 | 1254 | 113 | |
| GenBank mRNA | BC058679.1 | - | 8 - 9 | 1215 | 6834623 | 113 |
| M83336.1 | - | 7 - 8 | 1059 | 113 | ||
| X62646.1 | - | 7 - 8 | 1059 | 113 | ||
More Information
Set Membership: |
|

Il6st
Genomic Map