Assay Details
| Gene Symbol: Ano6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253813.1 | NP_001240742.1 | 16 - 17 | 2086 | 95 | |
| NM_175344.4 | NP_780553.2 | 15 - 16 | 2023 | 95 | ||
| GenBank mRNA | AK089300.1 | - | 16 - 17 | 2159 | 95 | |
| AK159411.1 | - | 16 - 17 | 2035 | 95 | ||
| BC060732.1 | - | 15 - 16 | 2023 | 6409332 | 95 | |
More Information
Set Membership: |
|

Ano6
Genomic Map