Assay Details
| Gene Symbol: Ano6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253813.1 | NP_001240742.1 | 4 - 5 | 491 | 105 | |
| NM_175344.4 | NP_780553.2 | 3 - 4 | 428 | 105 | ||
| GenBank mRNA | AK089300.1 | - | 3 - 4 | 339 | 105 | |
| AK159411.1 | - | 4 - 5 | 442 | 105 | ||
| BC060732.1 | - | 3 - 4 | 428 | 6409332 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK089300.1 transcripts.
Set Membership: |
|

Ano6
Genomic Map