Assay Details
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081402.1 | NP_001074871.1 | 14 - 15 | 1590 | 77 | |
GenBank mRNA | AK159714.1 | - | 14 - 15 | 1572 | 77 | |
AK160933.1 | - | 14 - 15 | 1527 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
