Assay Details
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081402.1 | NP_001074871.1 | 15 - 16 | 1677 | 89 | |
GenBank mRNA | AK159714.1 | - | 15 - 16 | 1659 | 89 | |
AK160933.1 | - | 15 - 16 | 1614 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
