Assay Details
| Gene Symbol: Dhrs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177564.5 | NP_808232.2 | 4 - 5 | 685 | 88 | |
| GenBank mRNA | AK157041.1 | - | 4 - 5 | 670 | 88 | |
| BC022224.1 | - | 4 - 5 | 646 | 4219994 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhrs11
Genomic Map