Assay Details
| Gene Symbol: Dhrs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177564.5 | NP_808232.2 | 6 - 7 | 845 | 66 | |
| GenBank mRNA | AK157041.1 | - | 6 - 7 | 830 | 66 | |
| BC022224.1 | - | 6 - 7 | 806 | 4219994 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
metabolic process
oxidation-reduction process
molecular_function
oxidoreductase activity
pinocarveol dehydrogenase activity
chloral hydrate dehydrogenase activity
hydroxymethylmethylsilanediol oxidase activity
1-phenylethanol dehydrogenase activity
myrtenol dehydrogenase activity
cis-1,2-dihydroxy-1,2-dihydro-8-carboxynaphthalene dehydrogenase activity
3-hydroxy-4-methyloctanoyl-CoA dehydrogenase activity
2-hydroxy-4-isopropenylcyclohexane-1-carboxyl-CoA dehydrogenase activity
cis-9,10-dihydroanthracene-9,10-diol dehydrogenase activity
citronellol dehydrogenase activity
naphthyl-2-hydroxymethyl-succinyl-CoA dehydrogenase activity
2,4,4-trimethyl-1-pentanol dehydrogenase activity
2,4,4-trimethyl-3-hydroxypentanoyl-CoA dehydrogenase activity
1-hydroxy-4,4-dimethylpentan-3-one dehydrogenase activity
endosulfan diol dehydrogenase activity
endosulfan hydroxyether dehydrogenase activity
3-hydroxy-2-methylhexanoyl-CoA dehydrogenase activity
3-hydroxy-2,6-dimethyl-5-methylene-heptanoyl-CoA dehydrogenase activity
versicolorin reductase activity
ketoreductase activity
biological_process
metabolic process
oxidation-reduction process
molecular_function
oxidoreductase activity
pinocarveol dehydrogenase activity
chloral hydrate dehydrogenase activity
hydroxymethylmethylsilanediol oxidase activity
1-phenylethanol dehydrogenase activity
myrtenol dehydrogenase activity
cis-1,2-dihydroxy-1,2-dihydro-8-carboxynaphthalene dehydrogenase activity
3-hydroxy-4-methyloctanoyl-CoA dehydrogenase activity
2-hydroxy-4-isopropenylcyclohexane-1-carboxyl-CoA dehydrogenase activity
cis-9,10-dihydroanthracene-9,10-diol dehydrogenase activity
citronellol dehydrogenase activity
naphthyl-2-hydroxymethyl-succinyl-CoA dehydrogenase activity
2,4,4-trimethyl-1-pentanol dehydrogenase activity
2,4,4-trimethyl-3-hydroxypentanoyl-CoA dehydrogenase activity
1-hydroxy-4,4-dimethylpentan-3-one dehydrogenase activity
endosulfan diol dehydrogenase activity
endosulfan hydroxyether dehydrogenase activity
3-hydroxy-2-methylhexanoyl-CoA dehydrogenase activity
3-hydroxy-2,6-dimethyl-5-methylene-heptanoyl-CoA dehydrogenase activity
versicolorin reductase activity
ketoreductase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27979839 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27979842 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27979838 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27979841 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432851 | BC022224.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432851 | AK157041.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432851 | NM_177564.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432851 | BC022224.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432851 | AK157041.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P432851 | NM_177564.5 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 23085 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814931 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331662 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873602 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821945 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 6052 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127230 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432851 | BC022224.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432851 | AK157041.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432851 | NM_177564.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432851 | BC022224.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432851 | AK157041.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P432851 | NM_177564.5 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979801 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27979802 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127230 | - | - |
| Whole Mouse Genome G4122A | A_51_P432851 | BC022224.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432851 | AK157041.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432851 | NM_177564.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432851 | BC022224.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432851 | AK157041.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P432851 | NM_177564.5 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122872 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127230 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09115944 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09115942 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09115939 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09115945 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27979841 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27979842 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27979838 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27979839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27979775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09157109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09734511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09554581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09401373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09115941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09734510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09401384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09441506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09157118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09554586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09528219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09401382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09115934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09270656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27979844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09157108 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10389305 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389307 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389302 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389301 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389304 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389303 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389306 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425704_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10389300 | - | - |
| Mouse Genome 430 2.0 Array | 1425704_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425704_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192970_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192970_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100031282.mm.1 | NM_177564.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031282.mm.1 | NM_177564.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031282.mm.1 | TC1100003418.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031283.mm.1 | NM_177564.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031283.mm.1 | NM_177564.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031283.mm.1 | TC1100003418.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425704_at | - | - |
| Mouse Expression Set 430 | 1425704_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425704_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425704_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425704_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425704_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 308193 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3190091 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003190091 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2746830 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2746830 | - | - |
| mouse-6 v1.1 (Illumina) | scl39804.8.1_101-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3190091 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39804.8.1_101-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190091 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2746830 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2746830 | - | - |

Dhrs11
Genomic Map