Assay Details
Gene Symbol: Psen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008943.2 | NP_032969.1 | 8 - 9 | 1362 | 91 | |
GenBank mRNA | AF149111.1 | - | 6 - 7 | 874 | 91 | |
AK134232.1 | - | 8 - 9 | 1045 | 91 | ||
AK134615.1 | - | 8 - 9 | 1084 | 91 | ||
AK148101.1 | - | 8 - 9 | 1083 | 91 | ||
AK165984.1 | - | 8 - 9 | 1083 | 91 | ||
AK169963.1 | - | 4 - 5 | 489 | 91 | ||
BC030409.1 | - | 8 - 9 | 1037 | 5133302 | 91 | |
BC071233.1 | - | 8 - 9 | 1101 | 3673609 | 91 | |
L42177.1 | - | 8 - 9 | 1058 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
