Assay Details
| Gene Symbol: Nadk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040395.4 | NP_001035485.2 | 5 - 6 | 849 | 82 | |
| NM_001085410.2 | NP_001078879.1 | 5 - 6 | 849 | 82 | ||
| NM_001286253.1 | NP_001273182.1 | 5 - 6 | 849 | 82 | ||
| GenBank mRNA | AK003858.1 | - | 5 - 6 | 426 | 82 | |
| AK016988.1 | - | 5 - 6 | 415 | 82 | ||
| AK046666.1 | - | 5 - 6 | 850 | 82 | ||
| AK078318.1 | - | 5 - 6 | 452 | 82 | ||
| AK162601.1 | - | 5 - 6 | 423 | 82 | ||
| BC115776.1 | - | 5 - 6 | 428 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nadk2
Genomic Map