Assay Details
| Gene Symbol: Nadk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040395.4 | NP_001035485.2 | 7 - 8 | 1065 | 94 | |
| NM_001286253.1 | NP_001273182.1 | 7 - 8 | 1065 | 94 | ||
| NM_001286255.1 | NP_001273184.1 | 6 - 7 | 981 | 94 | ||
| GenBank mRNA | AK003858.1 | - | 7 - 8 | 642 | 94 | |
| AK016988.1 | - | 7 - 8 | 631 | 94 | ||
| AK149267.1 | - | 6 - 7 | 561 | 94 | ||
| AK162601.1 | - | 7 - 8 | 639 | 94 | ||
| BC115776.1 | - | 7 - 8 | 644 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nadk2
Genomic Map