Assay Details
Gene Symbol: Nadk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040395.4 | NP_001035485.2 | 8 - 9 | 1160 | 62 | |
NM_001085410.2 | NP_001078879.1 | 9 - 10 | 1226 | 62 | ||
NM_001286253.1 | NP_001273182.1 | 8 - 9 | 1160 | 62 | ||
NM_001286255.1 | NP_001273184.1 | 7 - 8 | 1076 | 62 | ||
GenBank mRNA | AK003858.1 | - | 8 - 9 | 737 | 62 | |
AK016988.1 | - | 8 - 9 | 726 | 62 | ||
AK046666.1 | - | 9 - 10 | 1227 | 62 | ||
AK078318.1 | - | 9 - 10 | 829 | 62 | ||
AK149267.1 | - | 7 - 8 | 656 | 62 | ||
AK162601.1 | - | 8 - 9 | 734 | 62 | ||
BC115776.1 | - | 8 - 9 | 739 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
