Assay Details
Gene Symbol: Mid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011845.2 | NP_035975.1 | 3 - 4 | 886 | 78 | |
GenBank mRNA | AF196480.1 | - | 3 - 4 | 886 | 78 | |
Y18881.1 | - | 4 - 5 | 1241 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
