Assay Details
| Gene Symbol: C030037D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038058.1 | 2 - 3 | 544 | 80 | ||
| GenBank mRNA | AK021127.1 | - | 2 - 3 | 352 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | musashi RNA-binding protein 2 | Msi2 |
1700105C15Rik, AI451722, AI563628, AW489193, Msi2h |
-2.1kb | Anti-sense | Chr. 11 - 88339382 - 88718267 on NCBI Build GRCm38 |
| Downstream | A kinase (PRKA) anchor protein 1 | Akap1 |
AKAP121, AKAP84, Akap, C76494, C81186, DAKAP1, S-AKAP84 |
103.2kb | Anti-sense | Chr. 11 - 88830792 - 88864586 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

C030037D09Rik
Genomic Map