Assay Details
| Gene Symbol: Cd101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167906.1 | NP_001161378.1 | 2 - 3 | 425 | 82 | |
| GenBank mRNA | AM849328.1 | - | 2 - 3 | 425 | 82 | |
| AM849329.1 | - | 2 - 3 | 425 | 82 | ||
More Information
Set Membership: |
|

Cd101
Genomic Map