Assay Details
| Gene Symbol: Aldoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005077.1 | - | 3 - 4 | 586 | 80 | |
| AK039267.1 | - | 2 - 3 | 246 | 80 | ||
| BC004802.1 | - | 4 - 5 | 417 | 3589325 | 80 | |
| BC008184.1 | - | 4 - 5 | 417 | 3589205 | 80 | |
| S72537.1 | - | 4 - 5 | 441 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldoc
Genomic Map