Assay Details
| Gene Symbol: Aldoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005077.1 | - | 6 - 7 | 1000 | 97 | |
| AK039267.1 | - | 5 - 6 | 660 | 97 | ||
| BC004802.1 | - | 7 - 8 | 831 | 3589325 | 97 | |
| BC008184.1 | - | 7 - 8 | 831 | 3589205 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldoc
Genomic Map