Assay Details
| Gene Symbol: Aldoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ132391.1 | - | 1 - 2 | 160 | 59 | |
| AK005077.1 | - | 7 - 8 | 1204 | 59 | ||
| AK039267.1 | - | 6 - 7 | 864 | 59 | ||
| BC004802.1 | - | 8 - 9 | 1035 | 3589325 | 59 | |
| BC008184.1 | - | 8 - 9 | 1035 | 3589205 | 59 | |
| S72537.1 | - | 8 - 9 | 1059 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070992 | AK039267.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070992 | BC004802.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070992 | BC008184.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070992 | AJ132391.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2070992 | AK005077.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420681 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868421 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112684 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070992 | AK039267.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070992 | BC004802.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070992 | BC008184.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070992 | AJ132391.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2070992 | AK005077.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P220681 | S72537.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P220681 | AK039267.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P220681 | BC004802.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P220681 | BC008184.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P220681 | AJ132391.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P425284 | AK039267.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P425284 | AJ132391.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27942638 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112684 | - | - |
| Whole Mouse Genome G4122A | A_51_P425284 | AK039267.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P425284 | AJ132391.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P220681 | S72537.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P220681 | AK039267.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P220681 | BC004802.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P220681 | BC008184.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P220681 | AJ132391.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112684 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09618749 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09618763 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09618735 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09495061 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09618728 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09618731 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09618769 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09618747 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09618768 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09495062 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09495066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27942665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09618768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09618743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09495066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09618735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09618740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09495062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27942650 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10379156 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379158 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379162 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379160 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379161 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379157 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379159 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451461_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424714_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10379153 | - | - |
| Mouse Genome 430 2.0 Array | 1451461_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451461_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1424714_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424714_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11676_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11676_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100010759.mm.1 | NM_009657.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010759.mm.1 | TC1100001156.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100010760.mm.1 | NM_009657.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010760.mm.1 | TC1100001156.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451461_a_at | - | - |
| Mouse Expression Set 430 | 1424714_at | - | - |
| Mouse Expression Set 430 | 1451461_PM_a_at | - | - |
| Mouse Expression Set 430 | 1424714_PM_at | - | - |
| Murine Genome U74v2 Set | 160546_at | - | - |
| Murine Genome U74v2 Set | 160546_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451461_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424714_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451461_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424714_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424714_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451461_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424714_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451461_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 450121 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160137 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160137 | - | - |
| mouse-6 v1.1 (Illumina) | GI_60687505-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160137 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160137 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_60687505-S | - | - |

Aldoc
Genomic Map