Assay Details
| Gene Symbol: Foxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008238.2 | NP_032264.1 | 2 - 3 | 268 | 130 | |
| GenBank mRNA | BC108981.1 | - | 1 - 2 | 156 | 40047617 | 130 |
| X81593.1 | - | 2 - 3 | 218 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Foxn1
Genomic Map