Assay Details
| Gene Symbol: Foxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277290.1 | NP_001264219.1 | 6 - 7 | 1066 | 77 | |
| NM_008238.2 | NP_032264.1 | 6 - 7 | 1069 | 77 | ||
| GenBank mRNA | BC108980.1 | - | 5 - 6 | 954 | 40047616 | 77 |
| BC108981.1 | - | 5 - 6 | 957 | 40047617 | 77 | |
| X81593.1 | - | 6 - 7 | 1019 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Foxn1
Genomic Map