Assay Details
| Gene Symbol: Pigs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201406.1 | NP_958808.1 | 2 - 3 | 233 | 82 | |
| GenBank mRNA | AK033331.1 | - | 2 - 3 | 207 | 82 | |
| AK039604.1 | - | 2 - 3 | 204 | 82 | ||
| AK043232.1 | - | 2 - 3 | 207 | 82 | ||
| AK045759.1 | - | 2 - 3 | 204 | 82 | ||
| AK053771.1 | - | 2 - 3 | 207 | 82 | ||
| AK142871.1 | - | 2 - 3 | 204 | 82 | ||
| AK151858.1 | - | 2 - 3 | 195 | 82 | ||
| AK152229.1 | - | 2 - 3 | 195 | 82 | ||
| AK153300.1 | - | 2 - 3 | 199 | 82 | ||
| AK158128.1 | - | 2 - 3 | 204 | 82 | ||
| AK169633.1 | - | 2 - 3 | 199 | 82 | ||
| BC058979.1 | - | 2 - 3 | 233 | 5716747 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pigs
Genomic Map