Assay Details
| Gene Symbol: Morc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193309.1 | NP_001180238.1 | 8 - 9 | 1342 | 159 | |
| NM_029413.4 | NP_083689.2 | 8 - 9 | 1342 | 159 | ||
| GenBank mRNA | AK017472.1 | - | 8 - 9 | 1311 | 159 | |
| AK032807.1 | - | 8 - 9 | 1343 | 159 | ||
| BC036164.1 | - | 2 - 3 | 229 | 159 | ||
| BC098483.1 | - | 8 - 9 | 1297 | 30022418 | 159 | |
| BC132153.1 | - | 8 - 9 | 1196 | 40130430 | 159 | |
| BC132497.1 | - | 8 - 9 | 1196 | 40130774 | 159 | |
More Information
Updated Mapping Information
Set Membership: |
|

Morc4
Genomic Map