Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300008141.mm.1 |
NM_001281487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008141.mm.1 |
TC1300001278.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008143.mm.1 |
NM_001281487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008143.mm.1 |
TC1300001278.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008135.mm.1 |
NM_001281487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008135.mm.1 |
TC1300001278.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008140.mm.1 |
NM_001281487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008140.mm.1 |
TC1300001278.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008144.mm.1 |
NM_001281487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008144.mm.1 |
TC1300001278.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008139.mm.1 |
NM_001281487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008139.mm.1 |
TC1300001278.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008134.mm.1 |
NM_001281487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008134.mm.1 |
TC1300001278.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300008142.mm.1 |
NM_001281487.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300008142.mm.1 |
TC1300001278.mm.1 |
MTA TC |