Assay Details
| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145432.3 | NP_663407.2 | 2 - 3 | 412 | 84 | |
| GenBank mRNA | AK012216.1 | - | 2 - 3 | 356 | 84 | |
| AK076219.1 | - | 2 - 3 | 540 | 84 | ||
| AK132494.1 | - | 2 - 3 | 347 | 84 | ||
| AK149059.1 | - | 2 - 3 | 206 | 84 | ||
| AK158323.1 | - | 2 - 3 | 542 | 84 | ||
| AK169901.1 | - | 2 - 3 | 84 | |||
| BC065092.1 | - | 2 - 3 | 338 | 6855410 | 84 | |
| DV657360.1 | - | 2 - 3 | 412 | 84 | ||
More Information
Set Membership: |
|

Heatr6
Genomic Map