Assay Details
| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145432.3 | NP_663407.2 | 5 - 6 | 781 | 116 | |
| GenBank mRNA | AK012216.1 | - | 5 - 6 | 725 | 116 | |
| AK076219.1 | - | 5 - 6 | 909 | 116 | ||
| AK132494.1 | - | 5 - 6 | 716 | 116 | ||
| AK149059.1 | - | 5 - 6 | 575 | 116 | ||
| AK158323.1 | - | 5 - 6 | 911 | 116 | ||
| BC065092.1 | - | 5 - 6 | 707 | 6855410 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Heatr6
Genomic Map