Assay Details
| Gene Symbol: Htatsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028242.2 | NP_082518.1 | 3 - 4 | 619 | 101 | |
| NM_029371.1 | NP_083647.1 | 2 - 3 | 543 | 101 | ||
| GenBank mRNA | AK005527.1 | - | 2 - 3 | 479 | 101 | |
| AK011234.1 | - | 2 - 3 | 490 | 101 | ||
| AK012522.1 | - | 3 - 4 | 613 | 101 | ||
| AK049495.1 | - | 2 - 3 | 475 | 101 | ||
| BC037711.1 | - | 3 - 4 | 584 | 4221465 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Htatsf1
Genomic Map