Assay Details
| Gene Symbol: Htatsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028242.2 | NP_082518.1 | 4 - 5 | 669 | 105 | |
| NM_029371.1 | NP_083647.1 | 3 - 4 | 593 | 105 | ||
| GenBank mRNA | AK005527.1 | - | 3 - 4 | 529 | 105 | |
| AK011234.1 | - | 3 - 4 | 540 | 105 | ||
| AK012522.1 | - | 4 - 5 | 663 | 105 | ||
| AK049495.1 | - | 3 - 4 | 525 | 105 | ||
| AK139853.1 | - | 1 - 2 | 2194 | 105 | ||
| BC037711.1 | - | 4 - 5 | 634 | 4221465 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Htatsf1
Genomic Map