Assay Details
| Gene Symbol: Htatsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028242.2 | NP_082518.1 | 8 - 9 | 1178 | 58 | |
| NM_029371.1 | NP_083647.1 | 7 - 8 | 1102 | 58 | ||
| GenBank mRNA | AK011234.1 | - | 7 - 8 | 1049 | 58 | |
| AK049495.1 | - | 7 - 8 | 1034 | 58 | ||
| BC037711.1 | - | 8 - 9 | 1143 | 4221465 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Htatsf1
Genomic Map