Assay Details
Gene Symbol: Ddx26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172779.4 | NP_766367.1 | 11 - 12 | 1873 | 80 | |
GenBank mRNA | AK083942.1 | - | 11 - 12 | 1835 | 80 | |
AK144026.1 | - | 1 - 2 | 1718 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
