Assay Details
| Gene Symbol: Slc38a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178746.4 | NP_848861.1 | 9 - 10 | 1091 | 93 | |
| GenBank mRNA | AK037026.1 | - | 1 - 2 | 1423 | 93 | |
| AK043402.1 | - | 9 - 10 | 1079 | 93 | ||
| AK044735.1 | - | 9 - 10 | 1081 | 93 | ||
| AK078926.1 | - | 3 - 4 | 331 | 93 | ||
| BC052361.1 | - | 9 - 10 | 1091 | 6705789 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a9
Genomic Map