Assay Details
Gene Symbol: Srcin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018873.2 | NP_061361.2 | 4 - 5 | 585 | 63 | |
GenBank mRNA | AF040944.1 | - | 4 - 5 | 590 | 63 | |
U59873.1 | - | 4 - 5 | 589 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
