Assay Details
Gene Symbol: Larp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024526.2 | NP_001019697.2 | 5 - 6 | 706 | 103 | |
NM_001080948.2 | NP_001074417.1 | 5 - 6 | 719 | 103 | ||
NM_001284521.1 | NP_001271450.1 | 5 - 6 | 693 | 103 | ||
NM_001284522.1 | NP_001271451.1 | 5 - 6 | 716 | 103 | ||
NM_001284523.1 | NP_001271452.1 | 5 - 6 | 709 | 103 | ||
GenBank mRNA | AK049756.1 | - | 5 - 6 | 633 | 103 | |
AK172198.1 | - | 5 - 6 | 651 | 103 | ||
BC075726.1 | - | 4 - 5 | 3438 | 103 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
