Assay Details
Gene Symbol: Larp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024526.2 | NP_001019697.2 | 6 - 7 | 811 | 126 | |
NM_001080948.2 | NP_001074417.1 | 6 - 7 | 824 | 126 | ||
NM_001284521.1 | NP_001271450.1 | 6 - 7 | 798 | 126 | ||
NM_001284522.1 | NP_001271451.1 | 6 - 7 | 821 | 126 | ||
NM_001284523.1 | NP_001271452.1 | 6 - 7 | 814 | 126 | ||
GenBank mRNA | AK049756.1 | - | 6 - 7 | 738 | 126 | |
AK139909.1 | - | 6 - 7 | 662 | 126 | ||
AK165392.1 | - | 6 - 7 | 759 | 126 | ||
CN724171.1 | - | 2 - 3 | 158 | 126 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
