Assay Details
Gene Symbol: Larp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024526.2 | NP_001019697.2 | 9 - 10 | 1195 | 80 | |
NM_001080948.2 | NP_001074417.1 | 9 - 10 | 1208 | 80 | ||
NM_001284521.1 | NP_001271450.1 | 9 - 10 | 1182 | 80 | ||
NM_001284522.1 | NP_001271451.1 | 9 - 10 | 1205 | 80 | ||
NM_001284523.1 | NP_001271452.1 | 9 - 10 | 1198 | 80 | ||
GenBank mRNA | AK139909.1 | - | 9 - 10 | 1046 | 80 | |
AK165392.1 | - | 9 - 10 | 1143 | 80 | ||
CN724171.1 | - | 5 - 6 | 542 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
